Learn More
Cayman Chemical 1,3-Didocosahexaenoyl Glycerol, Purity: ≥98%, CAS: 140670-42-4, 1 mg
SDP
Catalog No. 502938531
Encompass_Preferred
A diacylglycerol; has been used in the synthesis of triacylglycerols that contain DHA at their sn-1 and sn-3 positions
Catalog No. 50-293-8531 Supplier Cayman Chemical Supplier No. 270111 MG
A diacylglycerol; has been used in the synthesis of triacylglycerols that contain DHA at their sn-1 and sn-3 positions