Learn More
Cayman Chemical 1 3-DIeIcosenoyl Glycerol 1mg
SDP
Catalog No. 502934059
Encompass_Preferred
A diacylglyerol containing 11(Z)-eicosenoic acid at the sn-1 and sn-3
Catalog No. 50-293-4059 Supplier Cayman Chemical Supplier No. 267951 MG
A diacylglyerol containing 11(Z)-eicosenoic acid at the sn-1 and sn-3