Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diethoxy-2-propanol 98.0+%, TCI America™
SDP

Catalog No. D046010ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D046010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D046010ML Supplier TCI America Supplier No. D046010ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4043-59-8
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
MDL Number MFCD00026934
InChI Key WIHIUTUAHOZVLE-UHFFFAOYSA-N
Synonym 1,3-diethoxy-2-propanol, diethylin, glycerol 1,3-diethyl ether, 1,3-glycerine diethyl ether, glycerol 1,3-bis ethyl ether, glycerol alpha,gamma-diethyl ether, 2-propanol, 1,3-diethoxy, unii-2g6n5234uj, 2-propanol,1,3-diethoxy, glycerol .alpha.,.gamma.-diethyl ether
PubChem CID 77664
IUPAC Name 1,3-diethoxypropan-2-ol
SMILES CCOCC(COCC)O

Specifications

Color Colorless
Boiling Point 191°C
Quantity 10 mL
UN Number 1993
Formula Weight 148.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diethoxy-2-propanol
RTECSNumber UB1476750
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.