Learn More
Medchemexpress LLC 1,3-diethyl 2-oxopropanedioate | 609-09-6 | MFCD00009121 | 95.0% | 174.15 g/mol | C7H10O5 | 50 G
SDP
Catalog No. 5000299868
Encompass_Preferred
1,3-Diethyl 2-oxopropanedioate (diethyl mesoxalate) is the diethyl ester of mesoxalic acid and is used as a reagent and intermediate in synthetic organic chemistry. It is employed in carbon-carbon bond-forming reactions and as a building block for small-molecule synthesis in research settings.
  • Used as an electrophilic building block in organic synthesis.
  • Suitable for small-molecule and medicinal chemistry applications.
  • Typically supplied at reagent-grade purity for laboratory use.
  • Available in laboratory-scale pack sizes for synthetic work.
Catalog No. 50-002-99868 Supplier Medchemexpress LLC Supplier No. HYW01680250G
1,3-Diethyl 2-oxopropanedioate (diethyl mesoxalate) is the diethyl ester of mesoxalic acid and is used as a reagent and intermediate in synthetic organic chemistry. It is employed in carbon-carbon bond-forming reactions and as a building block for small-molecule synthesis in research settings.
  • Used as an electrophilic building block in organic synthesis.
  • Suitable for small-molecule and medicinal chemistry applications.
  • Typically supplied at reagent-grade purity for laboratory use.
  • Available in laboratory-scale pack sizes for synthetic work.