Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diethylbenzene, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019304 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9304 5 mL
11-101-8771 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9304 Supplier MilliporeSigma Supelco Supplier No. 978655MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Diethylbenzenes are aromatic hydrocarbons that are classified under the class of volatile organic compounds (VOCs).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00009260

Specifications

CAS 141-93-5
Density 0.864 g/mL (at 20°C)
Flash Point 51°C - Closed Cup
Linear Formula C10H14
Quantity 5 mL
Molecular Weight (g/mol) 134.22
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 1,3-Diethylbenzene
Show More Show Less
RTECSNumber CZ5620000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.