Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Difluoro-2-propanol, 98%, Thermo Scientific™

Catalog No. AC175560100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC175560100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC175560100 Supplier Thermo Scientific Chemicals Supplier No. 175560100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 453-13-4
Molecular Formula C3H6F2O
Molecular Weight (g/mol) 96.08
MDL Number MFCD00000452
InChI Key PVDLUGWWIOGCNH-UHFFFAOYSA-N
Synonym 1,3-difluoro-2-propanol, 2-propanol, 1,3-difluoro, 1,3-difluoroisopropanol, 2-propanol,3-difluoro, wln: f1yq1f, sgqdlibrbrwup@, acmc-20ap4b, 1,3-difluoropropane-2-ol, 1,3-difluoro-propan-2-ol, 1,3-difluoro-2-hydroxypropane
PubChem CID 67985
IUPAC Name 1,3-difluoropropan-2-ol
SMILES OC(CF)CF

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown to Yellow
Density 1.2400g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.3720 to 1.3750 (20°C, 589nm)
Linear Formula FCH2CH(OH)CH2F
Quantity 10 g
Formula Weight 96.08
Percent Purity ≥97.5% (GC)
Physical Form Liquid
Chemical Name or Material 1, 3-Difluoro-2-propanol, 99%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-216-6
RTECSNumber UB1770000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.