Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Difluoro-5-iodobenzene, 98%

Catalog No. AAL1951003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1951003 1 g
AAL1951006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1951003 Supplier Thermo Scientific Chemicals Supplier No. L1951003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2265-91-0 | C6H3F2I | 239.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2265-91-0
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.99
MDL Number MFCD03094155
InChI Key QQCFOQNFPIAENW-UHFFFAOYSA-N
Synonym 3,5-difluoroiodobenzene, benzene, 1,3-difluoro-5-iodo, 1,3-difluoro-5-iodo-benzene, 3,5-difluoro iodobenzene, pubchem17134, 3,5-difluoro-iodobenzene, 3-difluoro-5-iodobenzene
PubChem CID 2778221
IUPAC Name 1,3-difluoro-5-iodobenzene
SMILES FC1=CC(I)=CC(F)=C1

Specifications

Boiling Point 173°C to 174°C
Flash Point 82°C (179°F)
Refractive Index 1.554
Quantity 1 g
Beilstein 2041185
Sensitivity Light sensitive
Formula Weight 239.99
Percent Purity 98%
Chemical Name or Material 1,3-Difluoro-5-iodobenzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.