Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[3-(difluoromethyl)-1,2-thiazol-5-yl]-3-[2-fluoro-5-(hydroxymethyl)-4-pyridinyl]urea | 2270879-17-7 | 98.9% | 50 MG
SDP

Catalog No. 5000231785
Encompass_Preferred

BRM/BRG1 ATP Inhibitor-1 is a potent, allosteric small-molecule inhibitor of the BRM (SMARCA2) and BRG1 (SMARCA4) ATPase activities used in epigenetic and cancer research.

  • Potent allosteric dual inhibitor of BRM and BRG1 ATPase activities (reported IC50 < 5 nM).
  • Molecular formula C11H9F3N4O2S; molecular weight 318.27 g/mol.
  • Typical purity ≥98% (example batch purity 98.86%), supplied as a solid powder.
  • Soluble in DMSO; suitable for cell-based and biochemical assays probing SMARCA2/4 function.

Catalog No. 50-002-31785 Supplier Medchemexpress LLC Supplier No. HY11937450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BRM/BRG1 ATP Inhibitor-1 is a potent, allosteric small-molecule inhibitor of the BRM (SMARCA2) and BRG1 (SMARCA4) ATPase activities used in epigenetic and cancer research.

  • Potent allosteric dual inhibitor of BRM and BRG1 ATPase activities (reported IC50 < 5 nM).
  • Molecular formula C11H9F3N4O2S; molecular weight 318.27 g/mol.
  • Typical purity ≥98% (example batch purity 98.86%), supplied as a solid powder.
  • Soluble in DMSO; suitable for cell-based and biochemical assays probing SMARCA2/4 function.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.