Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[3-(difluoromethyl)-1,2-thiazol-5-yl]-3-[2-fluoro-5-(hydroxymethyl)-4-pyridinyl]urea | 2270879-17-7 | 99.7% | 1 ML
SDP

Catalog No. 5000234159
Encompass_Preferred

BRM/BRG1 ATP Inhibitor-1 is an orally active, allosteric small-molecule inhibitor of the chromatin-remodeling ATPases BRM (SMARCA2) and BRG1 (SMARCA4), supplied for research use. It is offered as a white to off-white solid and as a 10 mM solution in DMSO (1 mL), and it shows low-nanomolar potency in biochemical assays (IC50 <5 nM).

  • Allosteric dual inhibitor of BRM and BRG1.
  • Molecular formula C11H9F3N4O2S, molecular weight ≈318.3 g/mol.
  • High reported purity for research applications.
  • Available as a solid and as a 10 mM DMSO solution (1 mL).
  • Recommended solvent: DMSO; prepare stock solutions and store at -80°C or -20°C per stability guidance.

Catalog No. 50-002-34159 Supplier Medchemexpress LLC Supplier No. HY11937410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BRM/BRG1 ATP Inhibitor-1 is an orally active, allosteric small-molecule inhibitor of the chromatin-remodeling ATPases BRM (SMARCA2) and BRG1 (SMARCA4), supplied for research use. It is offered as a white to off-white solid and as a 10 mM solution in DMSO (1 mL), and it shows low-nanomolar potency in biochemical assays (IC50 <5 nM).

  • Allosteric dual inhibitor of BRM and BRG1.
  • Molecular formula C11H9F3N4O2S, molecular weight ≈318.3 g/mol.
  • High reported purity for research applications.
  • Available as a solid and as a 10 mM DMSO solution (1 mL).
  • Recommended solvent: DMSO; prepare stock solutions and store at -80°C or -20°C per stability guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.