Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[3-(difluoromethyl)-1,2-thiazol-5-yl]-3-[2-fluoro-5-(hydroxymethyl)pyridin-4-yl]urea | 2270879-17-7 | 99.0% | 25 MG
SDP

Catalog No. 5000233904
Encompass_Preferred

Small-molecule inhibitor of BRM (SMARCA2) and BRG1 (SMARCA4) ATPase activity supplied as a milligram-scale research reagent for in vitro studies.

  • Targets BRM (SMARCA2) and BRG1 (SMARCA4) ATPase activity.
  • CAS registry number 2270879-17-7.
  • Chemical formula C11H9F3N4O2S.
  • Molecular weight 318.28 g/mol.
  • Reported purity up to 99% (supplier dependent).
  • Provided as a 25 mg powder, intended for research use only.

Catalog No. 50-002-33904 Supplier Medchemexpress LLC Supplier No. HY11937425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Small-molecule inhibitor of BRM (SMARCA2) and BRG1 (SMARCA4) ATPase activity supplied as a milligram-scale research reagent for in vitro studies.

  • Targets BRM (SMARCA2) and BRG1 (SMARCA4) ATPase activity.
  • CAS registry number 2270879-17-7.
  • Chemical formula C11H9F3N4O2S.
  • Molecular weight 318.28 g/mol.
  • Reported purity up to 99% (supplier dependent).
  • Provided as a 25 mg powder, intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.