Learn More
Medchemexpress LLC 1,3-dihydro-1-[2-hydroxy-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)-2H-benzimidazol-2-one | 153587-01-0 | MFCD00928190 | 98.1% | 362.23 | C15H8F6N2O2 | 50MG
SDP
Catalog No. 5000237310
Encompass_Preferred
NS-1619 is a selective activator (opener) of large-conductance, calcium-activated potassium (BK, KCa1.1) channels used as a research reagent to modulate membrane excitability in studies of vascular smooth muscle, neurons, and mitochondrial function. The compound is identified by CAS 153587-01-0, has molecular formula C15H8F6N2O2, and a molecular weight of 362.23 Da.
  • Selective BK channel activator for electrophysiology and pharmacology studies.
  • Applicable to research on vascular smooth muscle and neuronal excitability.
  • C hemical formula C15H8F6N2O2 and molecular weight 362.23 Da.
  • High purity (98.1% by LCMS) suitable for analytical and biological assays.
  • White to off-white solid form, appropriate for preparation of experimental stock solutions.
Catalog No. 50-002-37310 Supplier Medchemexpress LLC Supplier No. HY1249650MG
NS-1619 is a selective activator (opener) of large-conductance, calcium-activated potassium (BK, KCa1.1) channels used as a research reagent to modulate membrane excitability in studies of vascular smooth muscle, neurons, and mitochondrial function. The compound is identified by CAS 153587-01-0, has molecular formula C15H8F6N2O2, and a molecular weight of 362.23 Da.
  • Selective BK channel activator for electrophysiology and pharmacology studies.
  • Applicable to research on vascular smooth muscle and neuronal excitability.
  • C hemical formula C15H8F6N2O2 and molecular weight 362.23 Da.
  • High purity (98.1% by LCMS) suitable for analytical and biological assays.
  • White to off-white solid form, appropriate for preparation of experimental stock solutions.