Learn More
Medchemexpress LLC 1,3-dihydroxyacetone | 96-26-4 | MFCD00004670 | 98.0% | 90.08 g/mol | C3H6O3 | 1g
SDP
Catalog No. 5000268700
Encompass_Preferred
1 3-Dihydroxyacetone (DHA) the main active ingredient in sunless tanning skin-care preparations and an important precursor for the synthesis of various fine chemicals is produced on an industrial scale by microbial fermentation of glycerol (HY-B1659) in Gluconobacter oxydans 1 3-Dihydroxyacetone is also used for synthesis of new biodegradable polymers by combining with lactic acid (HY-B2227)[1][2]
Catalog No. 50-002-68700 Supplier Medchemexpress LLC Supplier No. HYY03351G
1 3-Dihydroxyacetone (DHA) the main active ingredient in sunless tanning skin-care preparations and an important precursor for the synthesis of various fine chemicals is produced on an industrial scale by microbial fermentation of glycerol (HY-B1659) in Gluconobacter oxydans 1 3-Dihydroxyacetone is also used for synthesis of new biodegradable polymers by combining with lactic acid (HY-B2227)[1][2]