Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dihydroxyacetone Dimer 98%, Thermo Scientific™

Catalog No. AC114795000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC114795000 500 g
AC114791000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC114795000 Supplier Thermo Scientific Chemicals Supplier No. 114795000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-26-4
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004670
InChI Key RXKJFZQQPQGTFL-UHFFFAOYSA-N
Synonym dihydroxyacetone, 1,3-dihydroxyacetone, chromelin, triulose, viticolor, dihyxal, oxantin, oxatone, soleal, otan
PubChem CID 670
ChEBI CHEBI:16016
IUPAC Name 1,3-dihydroxypropan-2-one
SMILES OCC(=O)CO

Specifications

Color White
Infrared Spectrum Authentic
Linear Formula C(=0)(CH2OH)2
Quantity 500 g
Merck Index 15, 3200
Formula Weight 90.08
Percent Purity ≥98.5% (on dry substance)
Physical Form Powder
Chemical Name or Material 1, 3-Dihydroxyacetone, 98%
Hazard Category HYGROSCOPIC
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.