Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Diiodo-5,5-Dimethylhydantoin

Catalog No. 7116245G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-162-45G 5 g
71-162-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 71-162-45G Supplier Sigma Aldrich Supplier No. 7116245G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DIH - Highly efficient and economical reagent for IodinationUsed for:Preparation of nitriles from corresponding alcohols and amines via oxidative conversionChemoselective, stereospecific iododesilylation of silylated alkenesPreparation of N-benzyl toluenesulfonamides via sulfonylamidation of alkylbenzenes with toluenesulfonamide in presence of diiododimethylhydantoinIodination reactions

Specifications

CAS 2232-12-4
Molecular Formula C5H6I2N2O2
Linear Formula C5H6I2N2O2
MDL Number MFCD00020867
Quantity 5 g
Synonym DIH
Molecular Weight (g/mol) 379.92 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.