Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethoxy-1,1,3,3-tetraphenyldisiloxane 90.0+%, TCI America™
SDP

Catalog No. D37345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37345G Supplier TCI America Supplier No. D37345G

Chemical Identifiers

CAS 94593-08-5
Molecular Formula C26H26O3Si2
Molecular Weight (g/mol) 442.66
MDL Number MFCD05663939
InChI Key CDHRQJOOOVAAFQ-UHFFFAOYSA-N
PubChem CID 21903287
IUPAC Name 3,3,5,5-tetraphenyl-2,4,6-trioxa-3,5-disilaheptane
SMILES CO[Si](O[Si](OC)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Boiling Point 210°C
Quantity 5 g
Formula Weight 442.66
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethoxy-1,1,3,3-tetraphenyldisiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.