Learn More
Medchemexpress LLC 1,3-Dimethoxybenzene | 151-10-0 | 99.99% | 138.17 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY3448710MM/1ML
Encompass_Preferred
1,3-Dimethoxybenzene is a volatile compound characterized by a sweet medicinal odor, featuring hazelnut, resinous, and woody notes. It has been identified as a new volatile component in Douro region grape musts and port wines.
  • Possesses a distinct sweet medicinal odor with notes of hazelnut, resin, and wood.
  • Identified as a new volatile component of Douro region grape musts and port wines.
  • Can be isolated from port wine extracts using flash chromatography.
Catalog No. 50-004-05058