Learn More
Medchemexpress LLC 1,3-Dimethoxybenzene | 151-10-0 | 99.99% | 138.17 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY34487S2100MG
Encompass_Preferred
1,3-Dimethoxybenzene is a volatile compound with a sweet, medicinal odor, carrying hazelnut, resinous, and woody notes. It has been identified as a new volatile component of Douro region grape musts and port wines.
  • Possesses sweet medicinal, hazelnut, resinous, and woody aromas.
  • Can be isolated from grape juice and Port wine.
  • Maximum content reaches up to 19.4 μg/L in 1999 vintage Port wine.
  • Primarily eluted in 60% hexane/40% diethyl ether fraction by flash chromatography.
Catalog No. 50-004-03723