Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Dimethoxybenzene | 151-10-0 | 99.99% | 138.17 | 25 MG
SDP

Catalog No. 5000402169
Encompass_Preferred

1,3-Dimethoxybenzene is a volatile compound with sweet medicinal, hazelnut, resinous, and woody notes. It has been identified as a new volatile component in grape musts and port wines from the Douro region. This product is intended for research use only.

  • Volatile compound with distinct aromas
  • Found in grape musts and port wines
  • Liquid appearance, colorless to light yellow
  • Ships at room temperature (continental US)
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: ≥ 100 mg/mL

Catalog No. 50-004-02169 Supplier Medchemexpress LLC Supplier No. 25MGHY34487S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dimethoxybenzene is a volatile compound with sweet medicinal, hazelnut, resinous, and woody notes. It has been identified as a new volatile component in grape musts and port wines from the Douro region. This product is intended for research use only.

  • Volatile compound with distinct aromas
  • Found in grape musts and port wines
  • Liquid appearance, colorless to light yellow
  • Ships at room temperature (continental US)
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.