Learn More
Medchemexpress LLC 1,3-Dimethoxybenzene | 151-10-0 | 99.99% | 138.17 | 50 G
SDP
Catalog No. 5000404234
Encompass_Preferred
1,3-Dimethoxybenzene is a volatile compound with a sweet medicinal odor, characterized by hazelnut, resinous, and woody notes. It has been identified as a new volatile component of Douro region grape musts and port wines. This compound can be isolated from grape juice and Port wine, with maximum content reaching up to 19.4 μg/L in 1999 vintage Port wine.
  • Volatile compound with a sweet medicinal odor
  • Features hazelnut, resinous, and woody notes
  • Identified as a component in Douro region grape musts and port wines
  • Can be isolated from grape juice and Port wine extracts
Catalog No. 50-004-04234 Supplier Medchemexpress LLC Supplier No. HY34487S50MG
1,3-Dimethoxybenzene is a volatile compound with a sweet medicinal odor, characterized by hazelnut, resinous, and woody notes. It has been identified as a new volatile component of Douro region grape musts and port wines. This compound can be isolated from grape juice and Port wine, with maximum content reaching up to 19.4 μg/L in 1999 vintage Port wine.
  • Volatile compound with a sweet medicinal odor
  • Features hazelnut, resinous, and woody notes
  • Identified as a component in Douro region grape musts and port wines
  • Can be isolated from grape juice and Port wine extracts