Learn More
Medchemexpress LLC 1,3-Dimethoxybenzene | 151-10-0 | 99.99% | 138.17 | 50 G
SDP
Supplier:  Medchemexpress LLC HY3448750G
Encompass_Preferred
1,3-Dimethoxybenzene is a volatile compound characterized by a sweet medicinal odor, featuring hazelnut, resinous, and woody notes. It has been identified as a new volatile component present in Douro region grape musts and port wines.
  • Is a volatile compound with a sweet medicinal odour carrying hazelnut, resinous, and woody notes.
  • Identified as a new volatile component of Douro region grape musts and port wines.
  • Possesses sweet medicinal, hazelnut, resinous and woody aromas.
  • Can be isolated from grape juice and port wine produced in the Douro River region.
  • Maximum content reaches up to 19.4 μg/L in 1999 vintage Port wine, 3 μg/L in 1998 vintage Port wine, 5.1 μg/L in 2000 vintage fortified Port wine, and 11 μg/L in 1998 vintage grape juice.
  • Can be isolated from port wine extracts by flash chromatography, primarily eluting in the 60% hexane/40% diethyl ether fraction.
Catalog No. 50-004-04569