Learn More
Medchemexpress LLC 1,3-Dimethoxybenzene | 151-10-0 | 99.99% | 138.17 | 500 G
SDP
Supplier:  Medchemexpress LLC HY34487500G
Encompass_Preferred
1,3-Dimethoxybenzene is a volatile compound characterized by a sweet medicinal odor with notes of hazelnut, resin, and wood. It has been identified as a new volatile component in grape musts from the Douro region and in port wines. This compound can be isolated from port wine extracts through flash chromatography, primarily eluting in the 60% hexane/40% diethyl ether fraction. The product is intended for research use only.
  • Volatile compound with a sweet medicinal odor carrying hazelnut, resinous, and woody notes.
  • Identified as a new volatile component of Douro region grape musts and port wines.
  • High purity of 99.99%.
Catalog No. 50-004-04549