Learn More
Medchemexpress LLC 1,3-Dimethoxybenzene | 151-10-0 | MFCD00008399 | 99.99% | 138.17 | 100 G
SDP
Supplier:  Medchemexpress LLC HY34487100G
Encompass_Preferred
1,3-Dimethoxybenzene is a volatile compound characterized by a sweet medicinal odor, along with hazelnut, resinous, and woody notes. It has been identified as a new volatile component found in grape musts and port wines from the Douro region. It can be isolated from grape juice and Port wine, with its maximum content reaching up to 19.4 μg/L in 1999 vintage Port wine.
  • Volatile compound with a sweet medicinal odor.
  • Features hazelnut, resinous, and woody notes.
  • Identified as a component in grape musts and port wines.
  • Can be isolated from port wine extracts.
Catalog No. 50-004-04548