Learn More
Medchemexpress LLC 1,3-dimethoxybenzene-d10 | 340257-57-0 | 148.23 | C8D10O2 | 10 MG
SDP
Catalog No. 5000297622
Encompass_Preferred
1,3-Dimethoxybenzene-d10 is the deuterium-labeled (d10) isotopologue of 1,3-dimethoxybenzene used as an internal standard and reagent in analytical and synthetic chemistry. It provides a predictable mass shift for mass spectrometry and distinct signals for NMR, and is supplied in small milligram quantities for laboratory workflows. This product is for research use only and not for human or clinical applications.
  • Deuterium-labeled isotopologue (d10) enabling mass-shifted detection in MS and distinct NMR signals.
  • Suitable as an internal standard for quantitative mass spectrometry methods.
  • Provided in small milligram pack sizes compatible with analytical and synthetic workflows.
  • Comes with datasheet and handling instructions; safety data sheet available on request.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-97622 Supplier Medchemexpress LLC Supplier No. HY34487S10MG
1,3-Dimethoxybenzene-d10 is the deuterium-labeled (d10) isotopologue of 1,3-dimethoxybenzene used as an internal standard and reagent in analytical and synthetic chemistry. It provides a predictable mass shift for mass spectrometry and distinct signals for NMR, and is supplied in small milligram quantities for laboratory workflows. This product is for research use only and not for human or clinical applications.
  • Deuterium-labeled isotopologue (d10) enabling mass-shifted detection in MS and distinct NMR signals.
  • Suitable as an internal standard for quantitative mass spectrometry methods.
  • Provided in small milligram pack sizes compatible with analytical and synthetic workflows.
  • Comes with datasheet and handling instructions; safety data sheet available on request.
  • Intended for research use only; not for human or clinical use.