Learn More
Medchemexpress LLC 1,3-Dimethoxybenzene-d10 | 340257-57-0 | 98% | 148.23 | 100 MG
SDP
Catalog No. 5000404233
Encompass_Preferred
1,3-Dimethoxybenzene-d10 is the deuterium labeled version of 1,3-Dimethoxybenzene. It belongs to the class of organic compounds known as dimethoxybenzenes and serves as an intermediate in the synthesis of organic compounds. This product is for research use only.
  • Deuterium-labeled compound.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Suitable for techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration can potentially influence drug pharmacokinetics and metabolism.
Catalog No. 50-004-04233 Supplier Medchemexpress LLC Supplier No. HY34487S100MG
1,3-Dimethoxybenzene-d10 is the deuterium labeled version of 1,3-Dimethoxybenzene. It belongs to the class of organic compounds known as dimethoxybenzenes and serves as an intermediate in the synthesis of organic compounds. This product is for research use only.
  • Deuterium-labeled compound.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Suitable for techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration can potentially influence drug pharmacokinetics and metabolism.