Learn More
Medchemexpress LLC 1,3-dimethoxybenzene-d6 | 151-10-0 | MFCD00008384 | 98.0% | 10 MG
SDP
Catalog No. 5000297865
Encompass_Preferred
1,3-dimethoxybenzene-d6 is the deuterated isotopologue of 1,3-dimethoxybenzene, provided as a research-scale labeled standard. Supplied at 98.0% purity in a 10 mg vial, it is intended for use in analytical workflows requiring deuterium labeling, such as mass spectrometry internal standards and tracer studies.
  • Deuterated isotopologue for accurate mass spectrometry and tracer studies.
  • High purity (98.0%) for reliable analytical performance.
  • Small, convenient 10 mg package for research-scale use.
  • Suitable as an internal standard for quantitative analysis.
  • Stable aromatic compound compatible with common organic solvents.
Catalog No. 50-002-97865 Supplier Medchemexpress LLC Supplier No. HY34487S210MG
1,3-dimethoxybenzene-d6 is the deuterated isotopologue of 1,3-dimethoxybenzene, provided as a research-scale labeled standard. Supplied at 98.0% purity in a 10 mg vial, it is intended for use in analytical workflows requiring deuterium labeling, such as mass spectrometry internal standards and tracer studies.
  • Deuterated isotopologue for accurate mass spectrometry and tracer studies.
  • High purity (98.0%) for reliable analytical performance.
  • Small, convenient 10 mg package for research-scale use.
  • Suitable as an internal standard for quantitative analysis.
  • Stable aromatic compound compatible with common organic solvents.