Learn More
Medchemexpress LLC 1,3-Dimethoxybenzene-d6 | 16469-85-5 | 98.0% | 144.20 | 25 MG
SDP
Catalog No. 5000402382
Encompass_Preferred
This product is 1,3-Dimethoxybenzene-d6, a deuterated compound suitable for laboratory use and substance manufacturing.
  • High isotopic enrichment of 99.9%
  • Purity of 98.0% (GC)
  • Suitable for laboratory research
  • Appears as a light yellow to yellow liquid
Catalog No. 50-004-02382 Supplier Medchemexpress LLC Supplier No. 25MGHY34487S2
This product is 1,3-Dimethoxybenzene-d6, a deuterated compound suitable for laboratory use and substance manufacturing.
  • High isotopic enrichment of 99.9%
  • Purity of 98.0% (GC)
  • Suitable for laboratory research
  • Appears as a light yellow to yellow liquid