Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Dimethoxybenzene-d6 | 16469-85-5 | 98.0% | 144.20 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY34487S25MG

Encompass_Preferred

1,3-Dimethoxybenzene-d6 is a laboratory chemical intended for the manufacture of substances and strictly for research use. It should only be handled by suitably qualified and experienced scientists in appropriately equipped and authorized facilities, as it has not been fully validated for medical applications. This product appears as a light yellow to yellow liquid.

  • Designed for research use only.
  • Not fully validated for medical applications.
  • Suitable for substance manufacturing.
  • Light yellow to yellow liquid appearance.
  • Store pure form at -20°C for up to 3 years.
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month.

Catalog No. 50-004-04570


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.