Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-2-(2-thienyl)imidazolidine, TCI America™
SDP

Catalog No. D22895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22895G Supplier TCI America Supplier No. D22895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104208-13-1
Molecular Formula C9H14N2S
Molecular Weight (g/mol) 182.285
MDL Number MFCD00191636
InChI Key HIHONCXGFIYIJX-UHFFFAOYSA-N
PubChem CID 2797481
IUPAC Name 1,3-dimethyl-2-thiophen-2-ylimidazolidine
SMILES CN1CCN(C1C2=CC=CS2)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 182.29
Physical Form Liquid
Chemical Name or Material 1,3-Dimethyl-2-(2-thienyl)imidazolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.