Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dimethyl-2-imidazolidinone

Catalog No. 40725100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-725-100ML 100 mL
40-725-500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-725-100ML Supplier Sigma Aldrich Supplier No. 40725100ML
Add to Cart
Add to Cart
This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,3-dimethyl-2-imidazolidinone (dmi) is an imidazolidine derivative. it is reported to act as a promoter by minimizing the formation of dialkylation byproducts and accelerating the rate of monoalkylation of α-butyrolactone. conversion of co2 to form lower alcohols by homogeneous catalytic hydrogenation using dmi as solvent has been investigated.1,3-dimethyl-2-imidazolidinone may be used:
  • As substitute solvent for hmpa in the synthesis of 1,2-bis(trimethylsilyl)benzene
  • As solvent during the α-regioselective prenylation of imines
  • A component of mobile phase during size-exclusion chromatographic analysis of cellulose.solvent used in various synthetic organic transformations. studied in the formation of functionally stabilized hydrosilanediyl-transition metal complexes produced photochemically from arylsilanes

Specifications

CAS 80-73-9
Density 1.056 g/mL (at 25°C (literature))
Boiling Point 224°C to 226°C (lit.)
Molecular Formula C5H10N2O
Refractive Index n20/D 1.472 (literature)
Linear Formula C5H10N2O
MDL Number MFCD00003188
Quantity 100 mL
Synonym N,N-Dimethylethyleneurea; DMEU; DMI
Molecular Weight (g/mol) 114.15
Percent Purity ≥99.5% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.