Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dimethyl-2-imidazolidinone

Catalog No. 193453100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-345-3100G 100 g
19-345-31KG 1 kg
19-345-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-345-3100G Supplier Sigma Aldrich Supplier No. 193453100G
Add to Cart
Add to Cart
This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,3-dimethyl-2-imidazolidinone constitutes the mobile phase during size-exclusion chromatographic analysis of cellulose.1,3-dimethyl-2-imidazolidinone was used as substitute solvent for toxic hmpa in the synthesis of 1,2-bis(trimethylsilyl)benzene
  • It was employed as solvent during α-regioselective prenylation of imines.solvent used in various synthetic organic transformations
  • Studied in the formation of functionally stabilized hydrosilanediyl-transition metal complexes produced photochemically from arylsilanes

Specifications

CAS 80-73-9
Density 1.056 g/mL (at 25°C (literature))
Boiling Point 224°C to 226°C (lit.)
Molecular Formula C5H10N2O
Refractive Index n20/D 1.472 (literature)
Linear Formula C5H10N2O
MDL Number MFCD00003188
Quantity 100 g
Synonym N,N-Dimethylethyleneurea; DMEU; DMI
Molecular Weight (g/mol) 114.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.