Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-2-imidazolidinone, Solstice

Catalog No. 6002818 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
600-28-18 1 L
600-28-17 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 600-28-18 Supplier Solstice Supplier No. 674841L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, GC-Headspace tested,≥99.5%

Solstice Chromasolv™ GC-headspace solvents:

  • developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • very pure solvents with extremely low concentrations of the defined residual solvents
  • state-of-the art production and application tests
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-73-9
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD00003188
InChI Key CYSGHNMQYZDMIA-UHFFFAOYSA-N
Synonym 1,3-dimethyl-2-imidazolidinone, n,n'-dimethylethyleneurea, dimethyl imidazolidinone, n,n'-dimethylimidazolidinone, 1,3-dimethylimidazolidone, rhonite 1, dmeu, karbomos tsem, 2-imidazolidinone, 1,3-dimethyl, 1,3-dimethylethyleneurea
PubChem CID 6661
IUPAC Name 1,3-dimethylimidazolidin-2-one
SMILES CN1CCN(C)C1=O

Specifications

Density 1.056g/mL at 25°C
Flash Point 95°C (203°F)
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 108808
Formula Weight 114.15g/mol
Percent Purity ≥99.5% (GC), ≥99.5%
Grade CHROMASOLV
Chemical Name or Material 1,3-Dimethyl-2-imidazolidinone
Hazard Statement P280-P305 + P351 + P338
RTECSNumber NJ0660000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.