Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dimethyl-3,4,5,6-Tetrahydro-2(1H)-Pyrimidinone

Catalog No. 251569500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-156-9500G 500 g
25-156-9100G 100 g
25-156-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 25-156-9500G Supplier Sigma Aldrich Supplier No. 251569500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Versatile solvent employed in N-alkylation of chiral, primary amines, O-alkylation of aldoses, and in the synthesis of poly(aryl ethers).

Specifications

CAS 7226-23-5
Density 1.06 g/mL at 25°C
Boiling Point 146°C (44 mmHg, lit.)
Molecular Formula C6H12N2O
Refractive Index n20/D 1.488
Linear Formula C6H12N2O
MDL Number MFCD00006550
Quantity 500 g
Synonym 1,3-Dimethyl-2-oxohexahydropyrimidine; N,N'-Dimethylpropylene urea; DMPU
Molecular Weight (g/mol) 128.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.