Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
1,3-Dimethylimidazolium-2-Carboxylate
Spécifications
Spécifications
| CAS | 922724-14-9 |
| Melting Point | 142°C to 146°C |
| Molecular Formula | C9H11NO |
| Linear Formula | C9H11NO |
| MDL Number | MFCD08275447 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 149.19 g/mol |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'