Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Dimethylpyrazole | 694-48-4 | 96.13 | 1 ML
SDP

Catalog No. 5000448866
Encompass_Preferred

1,3-Dimethylpyrazole is a bioactive compound isolated from Moso Bamboo Root, serving as an active small molecule and building block for the synthesis of multiple compounds. It is supplied as a colorless to light yellow liquid.

  • Molecular formula: C5H8N2
  • High purity (typically >99%)
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Handle in well-ventilated area, avoiding inhalation, skin, and eye contact
  • Wear appropriate personal protective equipment

Catalog No. 50-004-48866 Supplier Medchemexpress LLC Supplier No. HYW00216810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dimethylpyrazole is a bioactive compound isolated from Moso Bamboo Root, serving as an active small molecule and building block for the synthesis of multiple compounds. It is supplied as a colorless to light yellow liquid.

  • Molecular formula: C5H8N2
  • High purity (typically >99%)
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Handle in well-ventilated area, avoiding inhalation, skin, and eye contact
  • Wear appropriate personal protective equipment

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.