Learn More
Medchemexpress LLC 1,3-Dimethylpyrazole | 694-48-4 | 96.13 | 5 G
SDP
Catalog No. 5000448868
Encompass_Preferred
1,3-Dimethylpyrazole is a bioactive compound isolated from Moso Bamboo Root. It is an active small molecule that can be used as a building block for the synthesis of multiple compounds.
  • Bioactive compound isolated from Moso Bamboo Root
  • Active small molecule
  • Used as a building block for the synthesis of multiple compounds
Catalog No. 50-004-48868 Supplier Medchemexpress LLC Supplier No. HYW0021685G
1,3-Dimethylpyrazole is a bioactive compound isolated from Moso Bamboo Root. It is an active small molecule that can be used as a building block for the synthesis of multiple compounds.
  • Bioactive compound isolated from Moso Bamboo Root
  • Active small molecule
  • Used as a building block for the synthesis of multiple compounds