Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Dimethylpyrazole | 694-48-4 | MFCD00036665 | 96.13 | 50 G
SDP

Catalog No. 5000446951
Encompass_Preferred

1,3-Dimethylpyrazole is a bioactive compound isolated from Moso Bamboo Root. It is an active small molecule that can be used as a building block for the synthesis of various compounds.

  • Bioactive compound
  • Isolated from Moso Bamboo Root
  • Active small molecule
  • Building block for synthesis
  • Molecular weight: 96.13
  • Formula: C5H8N2
  • Purity: 99.8%
  • Appearance: Liquid
  • Color: Colorless to light yellow

Catalog No. 50-004-46951 Supplier Medchemexpress LLC Supplier No. HYW00216850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dimethylpyrazole is a bioactive compound isolated from Moso Bamboo Root. It is an active small molecule that can be used as a building block for the synthesis of various compounds.

  • Bioactive compound
  • Isolated from Moso Bamboo Root
  • Active small molecule
  • Building block for synthesis
  • Molecular weight: 96.13
  • Formula: C5H8N2
  • Purity: 99.8%
  • Appearance: Liquid
  • Color: Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.