Learn More
Selleck Chemical LLC 1 3-Dimethyluracil
SDP
Catalog No. 5000003012
Encompass_Preferred
1 3-Dimethyluracil (N N -dimethyluracil) is a methyl derivative of uric acid found occasionally in human urine
Catalog No. 50-000-03012 Supplier Selleck Chemical LLC Supplier No. S605025MG
Add to Cart
Add to Cart
1 3-Dimethyluracil (N N -dimethyluracil) is a methyl derivative of uric acid found occasionally in human urine