Learn More
Medchemexpress LLC 1,3-dimethyluric acid | 944-73-0 | MFCD00009740 | 99.4% | 196.16 g·mol⁻¹ | C7H8N4O3 | 100 MG
SDP
Catalog No. 5000302247
Encompass_Preferred
1,3-Dimethyluric acid is an oxopurine and a principal metabolic product of theophylline and caffeine. It is supplied as a high-purity solid for use as a reference standard and research reagent in drug metabolism, analytical method development, and biochemical studies of purine-related urinary calculi.
  • High purity suitable for analytical applications.
  • Reference standard for drug metabolism studies.
  • Solid form for convenient handling and storage.
  • Compatible with chromatographic and spectrometric assays.
  • Useful for studies of purine-derived urinary calculi.
Catalog No. 50-003-02247 Supplier Medchemexpress LLC Supplier No. HYW014993100MG
1,3-Dimethyluric acid is an oxopurine and a principal metabolic product of theophylline and caffeine. It is supplied as a high-purity solid for use as a reference standard and research reagent in drug metabolism, analytical method development, and biochemical studies of purine-related urinary calculi.
  • High purity suitable for analytical applications.
  • Reference standard for drug metabolism studies.
  • Solid form for convenient handling and storage.
  • Compatible with chromatographic and spectrometric assays.
  • Useful for studies of purine-derived urinary calculi.