Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-dimyristoyl-glycerol | 7770-09-4 | MFCD00048455 | 98.0% | 512.81 g/mol | C31H60O5 | 100 MG
SDP

Catalog No. 5000283591
Encompass_Preferred

1,3-Dimyristoyl-glycerol is a diacylglycerol lipid reagent used in life-science research for biochemical assays and lipid signaling studies. Supplied as a high-purity solid, it is suitable for analytical workflows and experimental applications.

  • Purity 98.0%.
  • Molecular formula C31H60O5.
  • Molecular weight 512.81 g/mol.
  • Available in quantities: 50 MG, 100 MG, 250 MG, 500 MG, 1 G.
  • Storage powder at -20°C for up to 3 years; in solvent -80°C for 6 months.
  • Intended for biochemical assays and lipid signaling research.

Catalog No. 50-002-83591 Supplier Medchemexpress LLC Supplier No. HYW127588100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dimyristoyl-glycerol is a diacylglycerol lipid reagent used in life-science research for biochemical assays and lipid signaling studies. Supplied as a high-purity solid, it is suitable for analytical workflows and experimental applications.

  • Purity 98.0%.
  • Molecular formula C31H60O5.
  • Molecular weight 512.81 g/mol.
  • Available in quantities: 50 MG, 100 MG, 250 MG, 500 MG, 1 G.
  • Storage powder at -20°C for up to 3 years; in solvent -80°C for 6 months.
  • Intended for biochemical assays and lipid signaling research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.