Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-dimyristoyl-glycerol | 7770-09-4 | MFCD00048455 | 98.0% | 512.81 g/mol | C31H60O5 | 50 MG
SDP

Catalog No. 5000282683
Encompass_Preferred

1,3-Dimyristoyl-glycerol is a diacylglycerol biochemical reagent used in lipid signaling and membrane biochemistry research. It is applied in biochemical assays to study diacylglycerol-mediated signaling pathways and protein kinase C activation. Supplied as a solid, it is handled and stored according to standard laboratory safety procedures.

  • High purity: 98.0%.
  • CAS number: 7770-09-4.
  • Chemical formula C31H60O5; molecular weight 512.81 g/mol.
  • Appearance: white to off-white solid.
  • Pack size: 50 mg.
  • Storage: powder stable at -20 °C; in solvent stable at -80 °C (6 months) or -20 °C (1 month).
  • Intended use: research reagent for biochemical and lipid studies.

Catalog No. 50-002-82683 Supplier Medchemexpress LLC Supplier No. HYW12758850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dimyristoyl-glycerol is a diacylglycerol biochemical reagent used in lipid signaling and membrane biochemistry research. It is applied in biochemical assays to study diacylglycerol-mediated signaling pathways and protein kinase C activation. Supplied as a solid, it is handled and stored according to standard laboratory safety procedures.

  • High purity: 98.0%.
  • CAS number: 7770-09-4.
  • Chemical formula C31H60O5; molecular weight 512.81 g/mol.
  • Appearance: white to off-white solid.
  • Pack size: 50 mg.
  • Storage: powder stable at -20 °C; in solvent stable at -80 °C (6 months) or -20 °C (1 month).
  • Intended use: research reagent for biochemical and lipid studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.