Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dinitrobenzene

Catalog No. NC2330996
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC2330996 100 g
501840303 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NC2330996 Supplier Sigma Aldrich Supplier No. D194255100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Dinitrobenzene
TRUSTED_SUSTAINABILITY

Specifications

CAS 99-65-0
Melting Point 84°C to 86°C (lit.)
Density 1.368 g/mL (at 25°C (literature))
Boiling Point 297°C (lit.)
Molecular Formula C6H4N2O4
Linear Formula C6H4(NO2)2
MDL Number MFCD00007222
Quantity 100 g
Molecular Weight (g/mol) 168.11
Percent Purity 97%anhydrous basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.