Learn More
Cayman Chemical 1 3-DInonadecnoyl Glycerol 1mg
SDP
Catalog No. 502932957
Encompass_Preferred
A diacylglycerol; has been used as a standard in the quantification of diacylglycerols in synthetic milk fat by SPE/HPLC/ELSD
Catalog No. 50-293-2957 Supplier Cayman Chemical Supplier No. 267701 MG
A diacylglycerol; has been used as a standard in the quantification of diacylglycerols in synthetic milk fat by SPE/HPLC/ELSD