Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Dioleoyl-2-palmitoylglycerol | 1716-07-0 | 859.39 | 1 ML
SDP

Catalog No. 5000417029
Encompass_Preferred

1,3-Dioleoyl-2-palmitoylglycerol is an important triacylglycerol used in infant formulas.

  • Purity: 99.97%
  • Appearance: liquid (Density: 0.9046 g/cm3)
  • Color: colorless to light yellow
  • Structure classification: lipid
  • Initial source: animals, human milk fats
  • Solubility (in vitro): soluble in DMSO (100 mg/mL, 116.36 mM; requires sonication; hygroscopic DMSO has significant impact on solubility)
  • Usage: for research use only. Not sold to patients.

Catalog No. 50-004-17029 Supplier Medchemexpress LLC Supplier No. HYN1037210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dioleoyl-2-palmitoylglycerol is an important triacylglycerol used in infant formulas.

  • Purity: 99.97%
  • Appearance: liquid (Density: 0.9046 g/cm3)
  • Color: colorless to light yellow
  • Structure classification: lipid
  • Initial source: animals, human milk fats
  • Solubility (in vitro): soluble in DMSO (100 mg/mL, 116.36 mM; requires sonication; hygroscopic DMSO has significant impact on solubility)
  • Usage: for research use only. Not sold to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.