Learn More
Medchemexpress LLC 1,3-dioleoyl-2-palmitoylglycerol | 1716-07-0 | MFCD00057803 | 100.0% | 859.39 | C55H102O6 | 10 MG
SDP
Catalog No. 5000296503
Encompass_Preferred
1,3-Dioleoyl-2-palmitoylglycerol is a triacylglycerol used as a lipid standard and research reagent, representative of triglycerides found in infant formula. It is supplied as a high-purity analytical standard suitable for biochemical studies and method development.
  • High purity: 99.97%.
  • Molecular formula C55H102O6 and molecular weight 859.39.
  • Available in solid and solution formats, including 10 mg and 10 mM (1 mL in DMSO).
  • Stored frozen for long-term stability; see storage guidelines for details.
  • For research use only; not for human consumption.
Catalog No. 50-002-96503 Supplier Medchemexpress LLC Supplier No. HYN1037210MG
1,3-Dioleoyl-2-palmitoylglycerol is a triacylglycerol used as a lipid standard and research reagent, representative of triglycerides found in infant formula. It is supplied as a high-purity analytical standard suitable for biochemical studies and method development.
  • High purity: 99.97%.
  • Molecular formula C55H102O6 and molecular weight 859.39.
  • Available in solid and solution formats, including 10 mg and 10 mM (1 mL in DMSO).
  • Stored frozen for long-term stability; see storage guidelines for details.
  • For research use only; not for human consumption.