Learn More
Cayman Chemical 1,3-Dioleoyl Glycerol, Purity: ≥98%, CAS: 2465-32-9, 10 mg
SDP
Catalog No. 502934753
Encompass_Preferred
A diacylglycerol; activates PKC at 50 μM
Catalog No. 50-293-4753 Supplier Cayman Chemical Supplier No. 2689610 MG
A diacylglycerol; activates PKC at 50 μM