Learn More
Cayman Chemical 1,3-Dioleoyl Glycerol, Purity: ≥98%, CAS: 2465-32-9, 250 mg
SDP
Catalog No. 502941469
Encompass_Preferred
A diacylglycerol; activates PKC at 50 μM
Catalog No. 50-294-1469 Supplier Cayman Chemical Supplier No. 26896250 MG
A diacylglycerol; activates PKC at 50 μM