Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-dioxo-2,3-dihydro-1H-isoindole-5-carboxylic a acid

Catalog No. CDS010434 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS010434 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS010434 Supplier Sigma Aldrich Supplier No. CDS01043425MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tert-butyl phenyl carbonate was used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography
  • It was also used in the synthesis of 2-nitroindoles
  • It was used as reagent for mono-boc protection of α,-d-diamines

Specifications

CAS 20262-55-9
Molecular Formula C9H5NO4
Linear Formula C9H5NO4
MDL Number MFCD00462255
Quantity 25 g
Molecular Weight (g/mol) 191.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.