Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1,3-Dioxolan-2-Ylmethyl)Magnesium Bromide Solution

Catalog No. 4726111L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-261-11L 1 L
47-261-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 47-261-11L Supplier Sigma Aldrich Supplier No. 4726111L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(1,3-Dioxolan-2-ylmethyl)magnesium bromide is a Grignard reagent that can be used to prepare:3-[11C]-Propionaldehyde, a key intermediate for the synthesis of radiolabeled compound [11C]SN-38, wherein SN-38 is 7-ethyl-10-hydroxy camptothecin.1-[(S)-2,2-Dimethyl-(1,3)-dioxolan-4-yl]-4-(1,3-dioxolan2-yl)-2-ol by reacting with D-glyceraldehyde, which is further used to synthesize unsaturated nucleosides.A bicyclo carboxylate intermediate, which is employed in the total synthesis of spirovibsanin A.

Specifications

CAS 180675-22-3
Density 0.938 g/mL at 25°C
Boiling Point 65°C to 67°C
Molecular Formula C4H7BrMgO2
Linear Formula C4H7BrMgO2
MDL Number MFCD01321271
Quantity 1 L
Molecular Weight (g/mol) 191.31 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.