Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1,3-Dioxolan-2-Ylmethyl)Zinc Bromide

Catalog No. 71032650ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-032-650ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-032-650ML Supplier Sigma Aldrich Supplier No. 71032650ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(1,3-Dioxolan-2-ylmethyl)zinc bromide is an organozinc compound used as a reagent in Negishi cross-coupling reaction to prepare aryl or heteroaryl scaffolds via C-C bond formation.(1,3-Dioxolan-2-ylmethyl)zinc bromide can be used as a reagent in the allylic alkylation reactions in the presence of iridium catalyst.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 947533-31-5
Melting Point 157°C to 161°C
Molecular Formula C8H13BN2O4
Linear Formula C8H13BN2O4
MDL Number MFCD09951915
Quantity 500 mg
Synonym 1-tert-Butoxycarbonyl-1H-pyrazole-4-boronic acid
Molecular Weight (g/mol) 212.01 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.