Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Dioxolane-4-ethanol, 2,2-di-(9Z,12Z)-9,12-octadecadien-1-yl- | 1190203-55-4 | C41H74O3 | 250 MG
SDP

Catalog No. 5000374380
Encompass_Preferred

LNP Lipid-7 is a cationic lipid used to prepare lipid nanoparticles (LNPs) for the delivery of drugs, such as nucleic acids.

  • Used to prepare lipid nanoparticles (LNPs) for drug delivery
  • Cationic lipid
  • Purity of 95.0%
  • Molecular weight of 615.02
  • Liquid appearance, colorless to light yellow
  • Density of 0.901 g/cm³

Catalog No. 50-003-74380 Supplier Medchemexpress LLC Supplier No. 250MGHY153852
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LNP Lipid-7 is a cationic lipid used to prepare lipid nanoparticles (LNPs) for the delivery of drugs, such as nucleic acids.

  • Used to prepare lipid nanoparticles (LNPs) for drug delivery
  • Cationic lipid
  • Purity of 95.0%
  • Molecular weight of 615.02
  • Liquid appearance, colorless to light yellow
  • Density of 0.901 g/cm³

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.